
4-(Trifluoromethyl)-2-pyridine carboxylic acid 97%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:588702-62-9
:
Formula:C₇H₄F₃NO₂
MDL Number:MFCD07438043
:191.11 g/mol
Melting Pt:157…162 °C
:GHS07
Storage Temperature:Ambient
Frequently Bought Together