

2-Fluoro-3-iodo-5-methylpyridine
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:153034-78-7
:
Formula:C₆H₅FIN
MDL Number:MFCD03095303
:237.02 g/mol
Flash Pt:>110 °C
:GHS05
:GHS07
Storage Temperature:Ambient
Frequently Bought Together