
1,6-Bis(acryloyloxy)-2,2,3,3,4,4,5,5-octafluorohexane 95%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:2264-01-9
:
Formula:C₁₂H₁₀F₈O₄
MDL Number:MFCD00078412
:370.20 g/mol
Density:1.2537 g/cm³ (25 °C)
Flash Pt:>110 °C
:GHS07
Storage Temperature:Ambient
Frequently Bought Together