4-{4-[3-Chloro-5-(trifluoromethyl)-2-pyridinyl]piperazino}aniline hydrochloride 97%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:321848-37-7
:
Formula:C₁₆H₁₇Cl₂F₃N₄
MDL Number:MFCD00793115
Melting Pt:230 °C
:393.24 g/mol
Storage Temperature:Ambient
Frequently Bought Together