[5-[(4-Chlorobenzyl)sulphanyl]-1-methyl-3-(trifluoromethyl)-1H-pyrazol-4-yl]methanol 97%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:318469-47-5
:
Formula:C₁₃H₁₂ClF₃N₂OS
MDL Number:MFCD00172667
:336.77 g/mol
Melting Pt:67…70 °C
Storage Temperature:Ambient
Frequently Bought Together