
3-Bromo-5-(trifluoromethyl)pyridin-2-ol 98%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:76041-73-1
:
Formula:C₆H₃BrF₃NO
MDL Number:MFCD02691223
:241.995 g/mol
Melting Pt:170…171 °C
:GHS07
Storage Temperature:Ambient
Frequently Bought Together