Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
£0.00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
United Kingdom
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Methyl-2,2,3,3-tetrafluoropropyl carbonate
Methyl-2,2,3,3-tetrafluoropropyl carbonate 99%
Methyl-2,2,3,3-tetrafluoropropyl carbonate 99%
APOSPC7886-1G
:
Apollo Scientific
:
Methyl-2,2,3,3-tetrafluoropropyl carbonate 99%
APOSPC7886-1G
:
Apollo Scientific
:
Pack type:
Bottle
Pk:
1 g
Seq:
3047
:
:
190.094 g/mol
:
GHS07
:
Ambient
:
156783-98-1
:
C₅H₆F₄O₃
:
MFCD08688580