

(2,4,5-Trifluorophenyl)acetic acid 97%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:209995-38-0
:
Formula:(C₆H₂F₃)CH₂COOH
MDL Number:MFCD00082479
:190.12 g/mol
Melting Pt:121…125 °C
:GHS05
:GHS07
Storage Temperature:Ambient
Frequently Bought Together