Methyl perfluoro-3,6-dioxadecanoate
Supplier: Apollo Scientific
Certificates
About this item
Synonyms: Methyl 2,2-difluoro-2-(1,1,2,2-tetrafluoro-2-(perfluorobutoxy)ethoxy)acetate
CAS Number:129301-40-2
:
Formula:C₉H₃F₁₅O₄
MDL Number:MFCD03094429
Boiling Pt:50 °C/12mmHg
Density:1.8 g/cm³ (25 °C)
Molecular Weight:460.10 g/mol
:GHS07
Storage Temperature:Ambient
Specifications
- Environmentally Preferable:
Specifications
Frequently Bought Together