Order Entry
United Kingdom
ContactUsLinkComponent
 
3,4,5-Trifluorobenzoyl chloride 97%
Catalog # APOSPC7285K-1G
Supplier:  Apollo Scientific
CAS Number:  
3,4,5-Trifluorobenzoyl chloride 97%
Catalog # APOSPC7285K-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :194.54 g/mol
Boiling Pt:173…174 °C
Storage Temperature:Ambient
UN:3265
ADR:8,II
 :GHS05
CAS Number:177787-26-7
Formula:C₇H₂ClF₃O
MDL Number:MFCD00083526