£0.00
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
United Kingdom
ContactUsLinkComponent
2,6-Difluoro-3-pyridineboronic acid hydrate
2,6-Difluoro-3-pyridineboronic acid hydrate
2,6-Difluoro-3-pyridineboronic acid hydrate
APOSPC7033-250MG
:
Apollo Scientific
:
2,6-Difluoro-3-pyridineboronic acid hydrate
APOSPC7033-250MG
:
Apollo Scientific
:
Pack type:
Bottle
Pk:
250 mg
Seq:
2590
Environmentally Preferable:
:
:
GHS07
:
Refrigerator
:
1072952-27-2
:
C₅H₄BF₂NO₂·H₂O
:
MFCD06657881