2,2,3,3-Tetrafluoro-1,4-butanediol 98%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:425-61-6
:
Formula:C₄H₆F₄O₂
MDL Number:MFCD00042375
:162.08 g/mol
Boiling Pt:110…112 °C/13mmHg
Melting Pt:80…87 °C
:GHS07
Storage Temperature:Ambient
Frequently Bought Together