4-Fluoro-1,3-dioxolan-2-one 97%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:114435-02-8
:
Formula:C₃H₃FO₃
MDL Number:MFCD06247543
:106.05 g/mol
Boiling Pt:212 °C
Melting Pt:28…23 °C
Flash Pt:102 °C
:GHS07
Storage Temperature:Refrigerator
Frequently Bought Together