
5-(Trifluoromethyl)-1,3,4-oxadiazol-2-amine
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:768-29-6
:
Formula:C₃H₂F₃N₃O
MDL Number:MFCD08062581
:153.06 g/mol
Melting Pt:155…160 °C
Storage Temperature:Refrigerator
UN:2811
ADR:6.1,III
:GHS06
Frequently Bought Together