Order Entry
United Kingdom
ContactUsLinkComponent
Perfluoro(methyldecalin) 85%
Perfluoro(methyldecalin) 85%
Catalog # APOSPC6120-25G
Supplier:  Apollo Scientific
CAS Number:  
Perfluoro(methyldecalin) 85%
Catalog # APOSPC6120-25G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    25 g
  • Seq:
    3308
  • Environmentally Preferable:

Specifications

About this item

Synonyms: Heptadecafluorodecahydro(trifluoromethyl)naphthalene, Perfluoro(methyldecahydronaphthalene)
 :
 :512.09 g/mol
 :GHS07
Storage Temperature:Ambient
CAS Number:306-92-3
 :51294-16-7
Formula:C₁₁F₂₀
MDL Number:MFCD00014326
Boiling Pt:160 °C
Density:1.96 g/cm³ (25 °C)