Order Entry
United Kingdom
ContactUsLinkComponent
2-Fluoro-6-pyridone 97%
2-Fluoro-6-pyridone 97%
Catalog # APOSPC5794-1G
Supplier:  Apollo Scientific
CAS Number:  
2-Fluoro-6-pyridone 97%
Catalog # APOSPC5794-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :113.09 g/mol
 :GHS07
Storage Temperature:Ambient
CAS Number:50543-23-2
Formula:C₅H₄FNO
MDL Number:MFCD03092938