Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
£0.00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
United Kingdom
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Perfluoro(allylbenzene)
Perfluoro(allylbenzene) 98%
Perfluoro(allylbenzene) 98%
Perfluoro(allylbenzene) 98%
APOSPC5697-100G
:
Apollo Scientific
:
Perfluoro(allylbenzene) 98%
APOSPC5697-100G
:
Apollo Scientific
:
Pack type:
Bottle
Pk:
100 g
Seq:
3401
Environmentally Preferable:
:
:
298.08 g/mol
:
GHS07
:
Ambient
:
67899-41-6
:
C₉F₁₀
:
MFCD00153225
:
136…138 °C
:
–38 °C
:
1.708 g/cm³ (25 °C)