Order Entry
United Kingdom
ContactUsLinkComponent
6-(Trifluoromethyl)pyridine-2-carbonyl chloride 97%
6-(Trifluoromethyl)pyridine-2-carbonyl chloride 97%
Catalog # APOSPC520906-1G
Supplier:  Apollo Scientific
CAS Number:  
6-(Trifluoromethyl)pyridine-2-carbonyl chloride 97%
Catalog # APOSPC520906-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :209.56 g/mol
Storage Temperature:Ambient
CAS Number:1099597-77-9
Formula:C₇H₃ClF₃NO
MDL Number:MFCD11226641