Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
£0.00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
United Kingdom
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
2-(Trifluoromethyl)benzyl mercaptan
2-(Trifluoromethyl)benzyl mercaptan 95%
2-(Trifluoromethyl)benzyl mercaptan 95%
APOSPC520547-5G
:
Apollo Scientific
:
2-(Trifluoromethyl)benzyl mercaptan 95%
APOSPC520547-5G
:
Apollo Scientific
:
Pack type:
Bottle
Pk:
5 g
Seq:
3173
:
:
Ambient
:
26039-98-5
:
C₈H₇F₃S