Methyl trifluoromethanesulphonate 98%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:333-27-7
:
Formula:C₂H₃F₃O₃S
MDL Number:MFCD00000409
:164.11 g/mol
Boiling Pt:94…102 °C
Melting Pt:–64 °C
Density:1.45 g/cm³ (25 °C)
Flash Pt:38 °C
Storage Temperature:Refrigerator
EINECS:206-371-7
UN:2920
ADR:8+3,II
:GHS02
:GHS05
Frequently Bought Together