
2'-Bromo-2,2,2-trifluoroacetophenone 97%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:244229-34-3
:
Formula:C₈H₄BrF₃O
MDL Number:MFCD07380806
Density:1.645 g/cm³ (25 °C)
Flash Pt:101 °C
:253.0160 g/mol
:GHS07
Storage Temperature:Ambient
Frequently Bought Together