Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
£0.00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
United Kingdom
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
5-Ethynyl-2-fluoropyridine
5-Ethynyl-2-fluoropyridine 97%
5-Ethynyl-2-fluoropyridine 97%
APOSPC49141-1G
:
Apollo Scientific
:
5-Ethynyl-2-fluoropyridine 97%
APOSPC49141-1G
:
Apollo Scientific
:
Pack type:
Bottle
Pk:
1 g
Seq:
3047
:
:
121.12 g/mol
:
Refrigerator
:
853909-08-7
:
C₇H₄FN
:
MFCD13175289