Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
£0.00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
United Kingdom
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
6-Fluoro-3-iodo-2-methylpyridine
6-Fluoro-3-iodo-2-methylpyridine 97%
6-Fluoro-3-iodo-2-methylpyridine 97%
6-Fluoro-3-iodo-2-methylpyridine 97%
APOSPC445047-1G
:
Apollo Scientific
:
6-Fluoro-3-iodo-2-methylpyridine 97%
APOSPC445047-1G
:
Apollo Scientific
:
Pack type:
Bottle
Pk:
1 g
Seq:
3047
:
:
237.02 g/mol
:
Ambient
:
884495-23-2
:
C₆H₅FIN
:
MFCD08277264