α,α,α,6-Tetrafluoro-o-toluic acid 98%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:32890-94-1
:
Formula:C₈H₄F₄O₂
MDL Number:MFCD00040981
:208.11 g/mol
Melting Pt:83…89 °C
:GHS07
Storage Temperature:Ambient
Frequently Bought Together