ω-Bromo-3-fluoroacetophenone 97%
Supplier: Apollo Scientific
Certificates
About this item
Synonyms: Bromomethyl m-fluorophenyl ketone, 3-Fluorophenacyl bromide, 2-Bromo-1-(3-fluorophenyl)ethan-1-one, α-Bromo-3-fluoroacetophenone, 2-Bromo-3'-fluoroacetophenone
CAS Number:53631-18-8
:
Formula:C₈H₆BrFO
MDL Number:MFCD00109756
Boiling Pt:76 °C/0.09mmHg
Molecular Weight:217.04 g/mol
Melting Pt:32…37 °C
Flash Pt:>110 °C
Storage Temperature:Refrigerator
UN:3261
ADR:8 III
:GHS05
Specifications
- Environmentally Preferable:
Specifications
Frequently Bought Together