Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
£0.00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
United Kingdom
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
3-Fluoro-2-trifluoromethylisonicotinic acid
3-Fluoro-2-trifluoromethylisonicotinic acid
3-Fluoro-2-trifluoromethylisonicotinic acid
APOSPC400548-1G
:
Apollo Scientific
:
3-Fluoro-2-trifluoromethylisonicotinic acid
APOSPC400548-1G
:
Apollo Scientific
:
Pack type:
Bottle
Pk:
1 g
Seq:
3047
:
:
209.1 g/mol
:
Ambient
:
886510-09-4
:
C₇H₃F₄NO₂
:
MFCD08437597