2,6-Difluoro-3-(propane-1-sulphonamido)benzoic acid 99%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:1103234-56-5
:
Formula:C₁₀H₁₁F₂NO₄S
MDL Number:MFCD18157658
:279.26 g/mol
Storage Temperature:Ambient
Frequently Bought Together