
4-Fluoro-2-methylanisole 97+%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:399-54-2
:
Formula:C₈H₉FO
MDL Number:MFCD00143319
:140.16 g/mol
Boiling Pt:69…70 °C/4mmHg
EINECS:609-758-2
:GHS07
Storage Temperature:Ambient
Frequently Bought Together