2-Bromo-N-cyclopropyl-4-(trifluoromethoxy)benzenesulphonamide
Catalog # APOSPC3494-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
Storage Temperature:Ambient
CAS Number:957062-79-2
Formula:C₁₀H₉BrF₃NO₃S
MDL Number:MFCD09878380