£0.00
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
United Kingdom
ContactUsLinkComponent
Ethyl perfluoro-n-dodecanoate
Ethyl perfluoro-n-dodecanoate 97%
Ethyl perfluoro-n-dodecanoate 97%
APOSPC3250M-1G
:
Apollo Scientific
:
Ethyl perfluoro-n-dodecanoate 97%
APOSPC3250M-1G
:
Apollo Scientific
:
Pack type:
Bottle
Pk:
1 g
Seq:
3047
Environmentally Preferable:
:
:
642.16 g/mol
:
GHS07
:
Ambient
:
158607-41-1
:
C₁₄H₅F₂₃O₂
:
MFCD00236676
:
96 °C/9mmHg