Order Entry
United Kingdom
ContactUsLinkComponent
 
Irritant
4-(Trifluoromethoxy)aniline hydrochloride
Catalog # APOSPC32088-5G
Supplier:  Apollo Scientific
CAS Number:  
Irritant
4-(Trifluoromethoxy)aniline hydrochloride
Catalog # APOSPC32088-5G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    5 g
  • Seq:
    3173

Specifications

About this item

 :
 :213.5900 g/mol
Storage Temperature:Ambient
UN:2811
ADR:6.1,III
 :GHS07
CAS Number:42823-24-5
Formula:C₇H₇ClF₃NO
MDL Number:MFCD00219956