4-Chloro-3-fluorobenzylamine 97%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:72235-58-6
:
Formula:C₇H₇ClFN
MDL Number:MFCD04116008
ADR:8,III
:159.59 g/mol
UN:2735
:GHS05
Storage Temperature:Ambient
Frequently Bought Together