3-Hydroxy-2-[3-(trifluoromethyl)phenyl]acrylonitrile 95%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:62739-04-2
:
Formula:C₁₀H₆F₃NO
MDL Number:MFCD12025877
:213.16 g/mol
Storage Temperature:Ambient
Frequently Bought Together