6-(4-Fluorophenyl)-2-oxo-1,2-dihydro-3-pyridinecarbonitrile 95%
Catalog # APOSPC27949-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:214.2 g/mol
Storage Temperature:Ambient
CAS Number:31755-80-3
Formula:C₁₂H₇FN₂O
MDL Number:MFCD00202736