2,3-Difluorobenzylamine 98%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:72235-51-9
:
Formula:C₇H₇F₂N
MDL Number:MFCD00070793
:143.14 g/mol
Boiling Pt:65 °C/4mmHg
Density:1.223 g/cm³ (25 °C)
Flash Pt:77 °C
Storage Temperature:Ambient
UN:2735
ADR:8,III
:GHS05
Frequently Bought Together