


3,5-Difluoroaniline 98%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:372-39-4
:
Formula:C₆H₅F₂N
MDL Number:MFCD00007763
:129.11 g/mol
Boiling Pt:80 °C/20mmHg
Melting Pt:40…42 °C
Flash Pt:75 °C
EINECS:206-752-8
UN:2811
ADR:6.1,III
:GHS06
:GHS07
:GHS08
Storage Temperature:Refrigerator
Frequently Bought Together