1,1-Dibromo-3,3,3-trifluoroacetone 95%
Supplier: Apollo Scientific
Certificates
About this item
Synonyms: 3,3-Dibromo-1,1,1-trifluoroacetone, 3,3-Dibromo-1,1,1-trifluoro-2-propanone, 3,3-Dibromo-1,1,1-trifluoropropan-2-one
CAS Number:431-67-4
:
Formula:Br₂HCC(O)CF₃
MDL Number:MFCD00041362
Boiling Pt:111…113 °C
Molecular Weight:269.84 g/mol
Storage Temperature:Refrigerator
UN:3265
ADR:8 III
:GHS05
Specifications
- Environmentally Preferable:
Specifications
Frequently Bought Together