

2-Chloro-4-(trifluoromethyl)pyrimidine-5-carbonyl chloride 97%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:154934-99-3
:
Formula:C₆HCl₂F₃N₂O
MDL Number:MFCD00173789
:244.99 g/mol
Boiling Pt:54 °C/0.15mmHg
UN:1760
ADR:8,III
:GHS05
:GHS07
Storage Temperature:Refrigerator
Frequently Bought Together