
2-Chloro-1,1,2-trifluoroethyl difluoromethyl ether 99+%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:13838-16-9
:
Formula:C₃H₂ClF₅O
MDL Number:MFCD00069095
Boiling Pt:56 °C
Density:1.517 g/cm³ (25 °C)
:184.49 g/mol
EINECS:207-553-4
:GHS07
Storage Temperature:Ambient
Frequently Bought Together