2-Chloro-3,4,4,4-tetrafluoro-3-(trifluoromethyl)-1-butene 97%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:235106-10-2
:
Formula:C₅H₂ClF₇
MDL Number:MFCD00155721
:230.51 g/mol
:GHS07
Storage Temperature:Ambient
Frequently Bought Together