Order Entry
United Kingdom
ContactUsLinkComponent
 
Methyl-3-bromo-2,6-difluorobenzoate
Catalog # APOSPC200499-500MG
Supplier:  Apollo Scientific
CAS Number:  
Methyl-3-bromo-2,6-difluorobenzoate
Catalog # APOSPC200499-500MG
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    500 mg
  • Seq:
    2725

Specifications

About this item

 :
 :251.03 g/mol
Storage Temperature:Ambient
CAS Number:1378875-92-3
Formula:C₈H₅BrF₂O₂
MDL Number:MFCD09951948