

(RS)-1-(3,5-Difluorophenyl)ethylamine 97%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:321318-29-0
:
Formula:C₈H₉F₂N
MDL Number:MFCD04038286
:157.16 g/mol
Boiling Pt:175 °C
UN:1760
ADR:8,III
:GHS05
:GHS07
Storage Temperature:Ambient
Frequently Bought Together