
5-Cyclopropyl-3-(trifluoromethyl)-1H-pyrazole
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:1027617-86-2
:
Formula:C₇H₇F₃N₂
MDL Number:MFCD03422575
:176.14 g/mol
Melting Pt:75…77 °C
:GHS07
Storage Temperature:Ambient
Frequently Bought Together