6-Fluoro-2-methylquinoline-3-carboxylic acid
Catalog # APOSPC1927-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:205.19 g/mol
Storage Temperature:Ambient
CAS Number:461026-47-1
Formula:C₁₁H₈FNO₂
MDL Number:MFCD03085774