


3-Chloro-2-fluoropyridine
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:1480-64-4
:
Formula:C₅H₃ClFN
MDL Number:MFCD04114144
:131.54 g/mol
Boiling Pt:162 °C
Density:1.33 g/cm³ (25 °C)
Flash Pt:52 °C
UN:2924
ADR:3+8,III
:GHS02
:GHS05
:GHS07
Storage Temperature:Ambient
Frequently Bought Together