Chlorodifluoroacetic anhydride 98%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:2834-23-3
:
Formula:C₄Cl₂F₄O₃
MDL Number:MFCD00000774
:242.94 g/mol
Boiling Pt:96…97 °C
Density:1.395 g/cm³ (25 °C)
Storage Temperature:Ambient
EINECS:220-603-4
UN:3265
ADR:8,II
:GHS05
:GHS07
Frequently Bought Together