5-Fluoro-3-methylsalicylaldehyde 98%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:704884-74-2
:
Formula:C₈H₇FO₂
MDL Number:MFCD04039244
:154.1384 g/mol
Boiling Pt:70…72 °C/5mmHg
Melting Pt:44…49 °C
Flash Pt:88 °C
:GHS05
:GHS07
Storage Temperature:Ambient
Frequently Bought Together