4-Bromo-3,3,4,4-tetrafluoro-1-butene 98%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:18599-22-9
:
Formula:C₄H₃BrF₄
MDL Number:MFCD00042189
:206.97 g/mol
Boiling Pt:55 °C
Density:1.627 g/cm³ (25 °C)
Flash Pt:8 °C
Storage Temperature:Refrigerator
EINECS:242-440-8
UN:1993
ADR:3,II
:GHS02
:GHS07
Frequently Bought Together