Order Entry
United Kingdom
ContactUsLinkComponent
 
Perfluoro(1-ethyl-3-propoxycyclohexane) 97%
  APOSPC1294-1G
 :  
Perfluoro(1-ethyl-3-propoxycyclohexane) 97%
  APOSPC1294-1G
 :  

 

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047
  • Environmentally Preferable:

 

 

 :
 :566.09 g/mol
 :Ambient
 :400626-82-6
 :C₁₁F₂₂O
 :MFCD03094121