Perfluoro(1,3-dipropoxycyclohexane) 97%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:400626-83-7
:
Formula:C₁₂F₂₄O₂
MDL Number:MFCD03094119
Molecular Weight:632.09 g/mol
Storage Temperature:Ambient
Specifications
- Environmentally Preferable:
Specifications
Frequently Bought Together